Få mere at vide
Olanzapine - d8, TRC
Artikelnummer. 30416562
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30416562 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416562 Leverandør Toronto Research Chemicals Leverandørnr. TRCO25379810MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Olanzapine-d8
CAS 1093380-13-2
Renhedsgrad noter HPLC
Molekylær formel C17 2H8 H12 N4 S
Mængde 10 mg
Synonym 10H-Thieno[2,3-b][1,5]benzodiazepine','2-methyl-4-(4-methyl-1-piperazinyl-2,2,3,3,5,5,6,6-d8)-, Olanzapine-D8
InChI formel InChI=1S/C17H20N4S/c1-12-11-13-16(21-9-7-20(2)8-10-21)18-14-5-3-4-6-15(14)19-17(13)22-12/h3-6,11,19H,7-10H2,1-2H3/i7D2,8D2,9D2,10D2
SMIL [2H]C1([2H])N(C)C([2H])([2H])C([2H])([2H])N(C2=Nc3ccccc3Nc4sc(C)cc24)C1([2H])[2H]
IUPAC navn 2-methyl-4-(2,2,3,3,5,5,6,6-octadeuterio-4-methylpiperazin-1-yl)-10H-thieno[2,3-b][1,5]benzodiazepine
Molekylvægt (g/mol) 320.48
Formel vægt 320.1911
Procent renhed >95