Få mere at vide
Olanzapine-d3 N-Oxide, TRC
Artikelnummer. 30427627
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30427627 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427627 Leverandør Toronto Research Chemicals Leverandørnr. TRCO25376210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Olanzapine-d3 N-Oxide
CAS 1189732-75-9
Molekylær formel C17H17D3N4OS
Mængde 10 mg
Synonym 2-Methyl-4-[4-(methyl-d3)-4-oxido-1-piperazinyl]-10H-thieno[2,3-b][1,5]benzodiazepine, LY 170238-d3
InChI formel InChI=1S/C17H20N4OS/c1-12-11-13-16(20-7-9-21(2,22)10-8-20)18-14-5-3-4-6-15(14)19-17(13)23-12/h3-6,11,19H,7-10H2,1-2H3/i2D3
SMIL CC1=CC2=C(S1)NC3=CC=CC=C3N=C2N4CCN(CC4)(C([2H])([2H])[2H])=O
Molekylvægt (g/mol) 331.45
Formel vægt 331.1546