Få mere at vide
Olanzapine-d3 (1.0mg/ml in Acetonitrile), TRC
Artikelnummer. 17999199
Skift visning
Mængde:
1 x 1 mL
Artikelnummer. Mængde
17999199 1 x 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 17999199 Leverandør Toronto Research Chemicals Leverandørnr. TRCKIT16351X1ML
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Olanzapine-methyl-D3
CAS 786686-79-1
Opløsningsmiddel eller Matrix Acetonitrile
Molekylær formel C17 2H3 H17 N4 S
Mængde 1 x 1 mL
Synonym 2-Methyl-4-(4-methyl-d3-1-piperazinyl)-10H-thieno[2,3-b][1,5]benzodiazepine, Integrol-d3, LY 170053-d3, Lanzac-d3, Oleanz-d3, Oliza-d3, Oltal-d3, Zyprexa-d3, 2-Methyl-4-[4-(methyl-d3)-1-piperazinyl]-10H-thieno[2,3-b][1,5]benzodiazepine (9CI), Olanzapine-D3
InChI formel InChI=1S/C17H20N4S/c1-12-11-13-16(21-9-7-20(2)8-10-21)18-14-5-3-4-6-15(14)19-17(13)22-12/h3-6,11,19H,7-10H2,1-2H3/i2D3
SMIL [2H]C([2H])([2H])N1CCN(CC1)C2=Nc3ccccc3Nc4sc(C)cc24
IUPAC navn 2-methyl-4-[4-(trideuteriomethyl)piperazin-1-yl]-10H-thieno[2,3-b][1,5]benzodiazepine
Molekylvægt (g/mol) 315.45
Formel vægt 315.1597