Learn More
Kern Sohn OIML Class M1 Beam Bar Test Weight
Beam bar weight constructed of brass.
POA - 925.62 DKK
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | ||||||
17302303
![]() |
Kern Sohn
347-445-100 |
10 g |
426.14 DKK
Each |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17312303
![]() |
Kern Sohn
347-445-100-N00 |
10 cN |
487.01 DKK
Each |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17382313
![]() |
Kern Sohn
347-475-100-N00 |
1 N | ||||||||
17392323
![]() |
Kern Sohn
347-495-100 |
500 g | ||||||||
17302333
![]() |
Kern Sohn
347-495-100-N00 |
5 N | ||||||||
17352333
![]() |
Kern Sohn
347-515-100 |
1000 g | ||||||||
17362333
![]() |
Kern Sohn
347-515-100-N00 |
10 N | ||||||||
Description
- Supporting bar shape
- Finely turned brass
Specifications
M1 | |
Brass |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Kern Sohn OIML Class M1 Beam Bar Test Weight