Få mere at vide
Tocris Bioscience™ Ogerin
Selective positive allosteric modulator at GPR68
Brand: Tocris Bioscience™ 5722/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Ogerin is a selective positive allosteric modulator (PAM) of GPR68 (pEC50 = 6.83), which exhibits selectivity for GPR68 over 58 other GPCRs. Ogerin attenuates contextual-based fear memory in mice.Tekniske data
| 2-[4-Amino-6-[(phenylmethyl)amino]-1,3,5-triazin-2-yl]benzenemethanol | |
| 1309198-71-7 | |
| 50 mg |