Få mere at vide
Ofloxacin Isopropyl Ester, TRC
Artikelnummer. 30424288
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424288 500 mg
30420063 1 g
30409729 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30424288 Leverandør Toronto Research Chemicals Leverandørnr. TRCO337025500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 126911-77-1
Molekylær formel C21 H26 F N3 O4
Molekylvægt (g/mol) 403.45
Synonym 9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid 1-methylethyl ester, 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, 9-fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-, 1-methylethyl ester, (±)-
IUPAC navn propan-2-yl 7-fluoro-2-methyl-6-(4-methylpiperazin-1-yl)-10-oxo-4-oxa-1-azatricyclo[7.3.1.05,13]trideca-5(13),6,8,11-tetraene-11-carboxylate
SMIL CC(C)OC(=O)C1=CN2C(C)COc3c(N4CCN(C)CC4)c(F)cc(C1=O)c23
Tekniske data
Kemisk navn eller materiale Ofloxacin Isopropyl Ester
Mængde 500 mg
InChI formel InChI=1S/C21H26FN3O4/c1-12(2)29-21(27)15-10-25-13(3)11-28-20-17(25)14(19(15)26)9-16(22)18(20)24-7-5-23(4)6-8-24/h9-10,12-13H,5-8,11H2,1-4H3
Formel vægt 403.1907