missing translation for 'onlineSavingsMsg'
Learn More

ODQ, 98%, Thermo Scientific Chemicals

Catalog Number p-3779366
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
41443-28-1
C9H5N3O2
187.2
MFCD00792620
LZMHWZHOZLVYDL-UHFFFAOYSA-N
1h-1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, odq, 1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, unii-s57v2nmv38, 1h-1,2,4 oxadiazolo 4,3-a-quinoxalin-1-one, 1h-1,2,4 oxadiazolo-4,3,a quinoxalin-1-one, 1-h-1,2,4 oxadiazolo-4,3,2 quinoxalin-1-ine, 1h-odq
1456
[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one
C1=CC=C2C(=C1)N=CC3=NOC(=O)N23
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
41443-28-1
C9H5N3O2
187.2
MFCD00792620
LZMHWZHOZLVYDL-UHFFFAOYSA-N
1h-1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, odq, 1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, unii-s57v2nmv38, 1h-1,2,4 oxadiazolo 4,3-a-quinoxalin-1-one, 1h-1,2,4 oxadiazolo-4,3,a quinoxalin-1-one, 1-h-1,2,4 oxadiazolo-4,3,2 quinoxalin-1-ine, 1h-odq
1456
[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one
C1=CC=C2C(=C1)N=CC3=NOC(=O)N23

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 160°C to 170°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 mg
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in chloroform,ethylacetate,dmso
Formula Weight 187.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material ODQ
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'