Learn More
Macherey-Nagel™ Octyl Phase Polygosil™ C8
Brand: Macherey-Nagel™ 711310.100
Additional Details : Weight : 0.01000kg
Specifications
7 μm | |
Liquid Chromatography | |
Polygosil C8 Phases |
60 Å | |
100 g |