missing translation for 'onlineSavingsMsg'
Få mere at vide

Octyl D-Galactofuranoside, TRC

Artikelnummer. 30426912
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426912 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30426912 Leverandør Toronto Research Chemicals Leverandørnr. TRCO241180250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Octyl D-Galactofuranoside
CAS 202403-49-4
Molekylær formel C14H28O6
Mængde 250 mg
InChI formel InChI=1S/C14H28O6/c1-2-3-4-5-6-7-8-19-14-12(18)11(17)13(20-14)10(16)9-15/h10-18H,2-9H2,1H3/t10?,11-,12?,13-,14?/m0/s1
SMIL CCCCCCCCOC1C([C@@H]([C@@H](O1)C(CO)O)O)O
IUPAC navn (2S,3S)-2-(1,2-dihydroxyethyl)-5-octoxyoxolane-3,4-diol
Molekylvægt (g/mol) 292.37
Formel vægt 292.1886
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.