Få mere at vide
Octyl beta-D-Mannopyranoside, TRC
Artikelnummer. 30423158
Skift visning
Mængde:
2.5 mg
10 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30423158 10 mg -
30414415 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30423158 Leverandør Toronto Research Chemicals Leverandørnr. TRCO29060010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Octyl beta-D-Mannopyranoside
CAS 140147-38-2
Molekylær formel C14 H28 O6
Mængde 10 mg
Synonym β-D-Mannopyranoside','octyl (9CI','ACI), Octyl β-D-mannopyranoside (ACI), 1-O-Octyl-β-D-mannopyranoside
InChI formel InChI=1S/C14H28O6/c1-2-3-4-5-6-7-8-19-14-13(18)12(17)11(16)10(9-15)20-14/h10-18H,2-9H2,1H3/t10-,11-,12+,13+,14-/m1/s1
SMIL CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]1O
IUPAC navn (2R,3S,4S,5S,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol
Molekylvægt (g/mol) 292.37
Formel vægt 292.1886