Få mere at vide
Octyl 2-(Acetylamino)-2-deoxy-Beta-D-glucopyranoside, TRC
Artikelnummer. 30417226
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30417226 100 mg
30424694 25 mg
30415642 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30417226 Leverandør Toronto Research Chemicals Leverandørnr. TRCO268050100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 147126-58-7
Molekylær formel C16 H31 N O6
Molekylvægt (g/mol) 333.42
IUPAC navn N-[(2R,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-octoxyoxan-3-yl]acetamide
SMIL CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1NC(=O)C
Tekniske data
Kemisk navn eller materiale Octyl 2-Acetamido-2-Deoxy-beta-D-Glucopyranoside
Mængde 100 mg
InChI formel InChI=1S/C16H31NO6/c1-3-4-5-6-7-8-9-22-16-13(17-11(2)19)15(21)14(20)12(10-18)23-16/h12-16,18,20-21H,3-10H2,1-2H3,(H,17,19)/t12-,13-,14-,15-,16-/m1/s1
Formel vægt 333.2151