Få mere at vide
MedChemExpress Octreotide
Tekniske data
Tekniske data
| Renhed | 98.84% |
| CAS | 83150-76-9 |
| Indhold og opbevaring | Protect from light∣Powder -80°C 2 years, -20°C 1 year∣The compound is unstable in solutions, freshly prepared is recommended. |
| Til brug med (applikation) | COVID-19-immunoregulation |
| Molekylvægt (g/mol) | 1019.24 g/mol |
| Produkttype | Peptides |
| Mængde | 10 mg |
| Forsendelsestilstand | Room temperature |
| Opløselighed | H2O : 100 mg/mL (98.11 mM∣Need ultrasonic) |
| Synonym | SMS 201-995 |