Learn More
Octinoxate, TRC
Product Code. 30009861
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009861 25 g 25g
30132582 100 g 100g
30135041 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009861 Supplier Toronto Research Chemicals Supplier No. TRCO25750025G
 Restricted Item
Forventet forsendelse: 04-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5466-77-3
Molecular Formula C18 H26 O3
Molecular Weight (g/mol) 290.4
Synonym 2-Ethylhexyl 4-methoxycinnamate, 2-Ethylhexyl methoxycinnamate, 2-Ethylhexyl p-methoxycinnamate, 3-(4-Methoxyphenyl)-2-propenoic acid 2-ethylhexyl ester, 4-Methoxycinnamic acid 2-ethylhexyl ester, Escalol 557, Escalol 557NB, Escalol 557T, Ethylhexyl methoxycinnamate, Ethylhexyl p-methoxycinnamate, Eusolex 2292, Eusolex UV-Pearls OMC, Jeescreen OMC, NSC 26466, Neo Heliopan AV, Nomcort TAB, Nomcort TAB-R, Octinoxate, Octyl 4-methoxycinnamate, Octyl methoxycinnamate, Octyl p-methoxycinnamate, Parsol MCX, Parsol MCX-SA, Parsol MOX, Solarom OMC, Sun Caps 664, Sunscreen AV, Tinosorb OMC, Uvinul 3088, Uvinul MC 80, Uvinul MC 80N, Uvinul MC 90, p-Methoxycinnamic acid 2-ethylhexyl ester
IUPAC Name 2-ethylhexyl 3-(4-methoxyphenyl)prop-2-enoate
SMILES CCCCC(CC)COC(=O)C=Cc1ccc(OC)cc1
Specifications
Quantity 25 g
InChI Formula InChI=1S/C18H26O3/c1-4-6-7-15(5-2)14-21-18(19)13-10-16-8-11-17(20-3)12-9-16/h8-13,15H,4-7,14H2,1-3H3
Formula Weight 290.1882
Percent Purity >95%
Chemical Name or Material Octinoxate
Grade HPLC