Få mere at vide
Octhilinone-d17, TRC
Artikelnummer. 30414032
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30414032 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414032 Leverandør Toronto Research Chemicals Leverandørnr. TRCO23930210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Octhilinone-d17
CAS 1185109-79-8
Renhedsgrad noter HPLC
Molekylær formel C11D17H2NOS
Mængde 10 mg
Synonym 2-Octyl-4-isothiazolin-3-one-d17, 2-Octyl-3(2H)-isothiazolone-d17, Pancil-D17, RH 893-d17, SD 888-d17, Skane 8-d17, Kathon-d17
InChI formel InChI=1S/C11H19NOS/c1-2-3-4-5-6-7-9-12-11(13)8-10-14-12/h8,10H,2-7,9H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,9D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])N1SC=CC1=O
IUPAC navn 2-(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecadeuteriooctyl)isothiazol-3-one
Molekylvægt (g/mol) 230.44
Formel vægt 230.2254
Procent renhed >95