Få mere at vide
Octaphonium Chloride, LoGiCal
Artikelnummer. 30457789
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30457789 5 mg
1 options
Artikelnummer. 30457789 Leverandør LoGiCal Leverandørnr. LGCFOR1966.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Octaphonium Chloride
CAS 15687-40-8
Analyt- eller komponentnavne Octaphonium Chloride
Forsendelsestilstand Room Temperature
Molekylær formel C27 H42 N O . Cl
Certificeringer/compliance ISO 9001
Synonym Benzenemethanaminium, N,N-diethyl-N-[2-[4-(2,2,4-trimethylpentyl)phenoxy]ethyl]-, chloride (1:1), Ammonium, benzyldiethyl[2-[p-(2,2,4-trimethylpentyl)phenoxy]ethyl]-, chloride (8CI), Benzenemethanaminium, N,N-diethyl-N-[2-[4-(2,2,4-trimethylpentyl)phenoxy]ethyl]-, chloride (9CI), Benzyldiethyl(2-[4-(2,2,4-trimethylpentyl)phenoxy]ethyl)ammonium chloride, Octafonium chloride, Octaphonium chloride
InChI formel InChI=1S/C27H42NO.ClH/c1-7-28(8-2,22-25-12-10-9-11-13-25)18-19-29-26-16-14-24(15-17-26)21-27(5,6)20-23(3)4;/h9-17,23H,7-8,18-22H2,1-6H3;1H/q+1;/p-1
Løsningstype Neat
SMIL [Cl-].CC[N+](CC)(CCOc1ccc(CC(C)(C)CC(C)C)cc1)Cc2ccccc2
IUPAC navn benzyl-diethyl-[2-[4-(2,2,4-trimethylpentyl)phenoxy]ethyl]azanium;chloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 432.08
Mængde 5 mg
Formel vægt 431.2955 g/mol