Få mere at vide
Octanoylamide Propylbetaine, TRC
Artikelnummer. 30136061
Skift visning
Mængde:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136061 2.5 g
30010645 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136061 Leverandør Toronto Research Chemicals Leverandørnr. TRCO2380152.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 73772-46-0
Molekylær formel C15H30N2O3
Molekylvægt (g/mol) 286.41
Synonym 2-(Dimethyl(3-octanamidopropyl)ammonio)acetate;
IUPAC navn 2-[dimethyl-[3-(octanoylamino)propyl]azaniumyl]acetate
SMIL CCCCCCCC(=O)NCCC[N+](C)(C)CC(=O)[O-]
Tekniske data
Kemisk navn eller materiale Octanoylamide Propylbetaine
Mængde 2.5 g
InChI formel InChI=1S/C15H30N2O3/c1-4-5-6-7-8-10-14(18)16-11-9-12-17(2,3)13-15(19)20/h4-13H2,1-3H3,(H-,16,18,19,20)
Formel vægt 286.2256