Få mere at vide
Octanoyl L-Carnitine-d3 Chloride, TRC
Artikelnummer. 17988749
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17988749 2.5 mg 2.5mg
17948979 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 17988749 Leverandør Toronto Research Chemicals Leverandørnr. TRCO2380072.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1334532-24-9
Molekylær formel C15 D3 H27 N O4 . Cl
Molekylvægt (g/mol) 326.87
Synonym Octanoyl-L-carnitine-d3 HCl (N-methyl-d3)
IUPAC navn [(2R)-3-carboxy-2-octanoyloxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride
SMIL [Cl-].[2H]C([2H])([2H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CCCCCCC
Tekniske data
Kemisk navn eller materiale Octanoyl L-Carnitine-d3 Chloride
Mængde 2.5 mg
InChI formel InChI=1S/C15H29NO4.ClH/c1-5-6-7-8-9-10-15(19)20-13(11-14(17)18)12-16(2,3)4;/h13H,5-12H2,1-4H3;1H/t13-;/m1./s1/i2D3;
Formel vægt 326.2052