Få mere at vide
Octanoic Acid-D15, TRC
Artikelnummer. 30414736
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30414736 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414736 Leverandør Toronto Research Chemicals Leverandørnr. TRCO2378821G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Octanoic acid-D15
CAS 69974-55-6
Renhedsgrad noter GC
Molekylær formel C8 2H15 H O2
Mængde 1 g
Synonym Octanoic-2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-d15 acid (ACI), Octanoic-d15 acid (9CI), 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecadeuteriooctanoic acid, Octanoic acid D15, Octanoic acid-d15, Caprylic acid-d15
InChI formel InChI=1S/C8H16O2/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3,(H,9,10)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecadeuteriooctanoic acid
Molekylvægt (g/mol) 159.3
Formel vægt 159.2092
Procent renhed >95