Få mere at vide
Octanohydroxamic Acid, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Octanohydroxamic acid |
| CAS | 7377-03-9 |
| Molekylær formel | C8 H17 N O2 |
| Mængde | 500 mg |
| Synonym | N-Hydroxyoctanamide, Caprylohydroxamic Acid, Decoyl Hydroxamic Acid, Octanoylhydroxamic Acid, Taselin |
| InChI formel | InChI=1S/C8H17NO2/c1-2-3-4-5-6-7-8(10)9-11/h11H,2-7H2,1H3,(H,9,10) |
| SMIL | CCCCCCCC(=O)NO |
| IUPAC navn | octanehydroxamic acid |
| Molekylvægt (g/mol) | 159.23 |
| Formel vægt | 159.1259 |