Få mere at vide
Octanal-d16, TRC
Artikelnummer. 30418829
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30418829 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418829 Leverandør Toronto Research Chemicals Leverandørnr. TRCO23720210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Octanal-d16
CAS 1219794-66-7
Molekylær formel C8 2H16 O
Mængde 10 mg
Synonym Octanal-d16, Octanal-1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-d16
InChI formel InChI=1S/C8H16O/c1-2-3-4-5-6-7-8-9/h8H,2-7H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D
SMIL [2H]C(=O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-hexadecadeuteriooctan-1-one
Molekylvægt (g/mol) 144.31
Formel vægt 144.2205