Få mere at vide
Octadecanoic-18,18,18-d3 Acid, TRC
Artikelnummer. 30426827
Skift visning
Mængde:
2.5 mg
5 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30426827 25 mg -
30419226 2.5 mg 2.5mg
30413040 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426827 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68649925MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 62163-39-7
Molekylær formel C18H33D3O2
Molekylvægt (g/mol) 287.5
Synonym 18,18,18-Trideuterostearic acid, [18,18,18-d3]-Stearic acid, Stearic acid-18,18,18-d3
IUPAC navn octadecanoic-18,18,18-d3 acid
SMIL OC(CCCCCCCCCCCCCCCCC([2H])([2H])[2H])=O
Tekniske data
Kemisk navn eller materiale Octadecanoic-18,18,18-D3 Acid
Mængde 25 mg
InChI formel InChI=1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i1D3
Formel vægt 287.29