Få mere at vide
Octadecanoic-18,18,18-d3 Acid, CDN
Artikelnummer. 30319610
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319610 0.25 g -
30316197 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319610 Leverandør CDN Leverandørnr. CDND18250.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Octadecanoic-18,18,18-D3 Acid
CAS 62163-39-7
Molekylær formel C18H33D3O2
Mængde 0.25 g
Synonym 18,18,18-Trideuterostearic acid, [18,18,18-d3]-Stearic acid, Stearic acid-18,18,18-d3
InChI formel InChI=1 S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i1D3
SMIL OC(CCCCCCCCCCCCCCCCC([2 H])([2 H])[2 H])=O
IUPAC navn octadecanoic-18,18,18-d3 acid
Molekylvægt (g/mol) 287.5
Formel vægt 287.29 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity