Få mere at vide
Octadecanoic-17,17,18,18,18-d5 Acid, TRC
Artikelnummer. 30418242
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30418242 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418242 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6865061MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Octadecanoic-17,17,18,18,18-D5 Acid
CAS 211443-83-3
Molekylær formel C18H31D5O2
Mængde 1 mg
Synonym Octadecanoic-17,17,18,18,18-d5 Acid, Octadecanoic-17,17,18,18,18-d5 acid (9CI), Stearic-17,17,18,18,18-d5 acid, 17,17,18,18,18-Pentadeuterostearic acid, [17,17,18,18,18-d5]-Stearic acid, Stearic acid-17,17,18,18,18-d5, Octadecanoic-17,17,18,18,18-D5 acid
InChI formel InChI=1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i1D3,2D2
SMIL OC(CCCCCCCCCCCCCCCC([2H])([2H])C([2H])([2H])[2H])=O
IUPAC navn octadecanoic-17,17,18,18,18-d5 acid
Molekylvægt (g/mol) 289.51
Formel vægt 289.3