missing translation for 'onlineSavingsMsg'
Learn More

Octacosane, 99+%, Thermo Scientific Chemicals

Catalog Number p-3737100
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
630-02-4
C28H58
394.77
MFCD00009355
ZYURHZPYMFLWSH-UHFFFAOYSA-N
n-octacosane, n-octcosane, unii-vff49836p8, ch3-ch2 26-ch3, octacosane, analytical standard, ccris 680, octacosane, n, octacosane, heptacosane, methyl, n-octacosane 10g
12408
CHEBI:32943
octacosane
CCCCCCCCCCCCCCCCCCCCCCCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
630-02-4
C28H58
394.77
MFCD00009355
ZYURHZPYMFLWSH-UHFFFAOYSA-N
n-octacosane, n-octcosane, unii-vff49836p8, ch3-ch2 26-ch3, octacosane, analytical standard, ccris 680, octacosane, n, octacosane, heptacosane, methyl, n-octacosane 10g
12408
CHEBI:32943
octacosane
CCCCCCCCCCCCCCCCCCCCCCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 61.0°C to 64.0°C
Color White
Density 0.8067g/mL
Boiling Point 440.0°C
Flash Point 227°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)26CH3
Quantity 5 g
Beilstein 01,I,71
Specific Gravity 0.8067
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, chloroform
Formula Weight 394.76
Percent Purity 99+%
Physical Form Shiny Flakes or Powder
Chemical Name or Material Octacosane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'