Få mere at vide
Octacaine Hydrochloride, LoGiCal
Artikelnummer. 30463068
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463068 10 mg
1 options
Artikelnummer. 30463068 Leverandør LoGiCal Leverandørnr. LGCFOR1921.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Octacaine Hydrochloride
CAS 93940-32-0
Analyt- eller komponentnavne Octacaine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C14 H22 N2 O . Cl H
Certificeringer/compliance ISO 9001
Synonym Butanamide, 3-(diethylamino)-N-phenyl-, hydrochloride (1:1), Butanamide, 3-(diethylamino)-N-phenyl-, monohydrochloride (9CI), Amplicain, Amplicain hydrochloride, Octacaine Hydrochloride
InChI formel InChI=1S/C14H22N2O.ClH/c1-4-16(5-2)12(3)11-14(17)15-13-9-7-6-8-10-13;/h6-10,12H,4-5,11H2,1-3H3,(H,15,17);1H
Løsningstype Neat
SMIL Cl.CCN(CC)C(C)CC(=O)Nc1ccccc1
IUPAC navn 3-(diethylamino)-N-phenylbutanamide;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 270.8
Mængde 10 mg
Formel vægt 270.1499 g/mol