Få mere at vide
Ochratoxin C-d5, TRC
Artikelnummer. 30421621
Skift visning
Mængde:
0.25 mg
Artikelnummer. Mængde
30421621 0.25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421621 Leverandør Toronto Research Chemicals Leverandørnr. TRCO1485120.25MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ochratoxin C-d5
CAS 1356840-94-2
Molekylær formel C22H17D5ClNO6
Mængde 0.25 mg
Synonym N-[[(3R)-5-Chloro-3,4-dihydro-8-hydroxy-3-methyl-1-oxo-1H-2-benzopyran-7-yl]carbonyl]-L-(phenyl-d5)alanine Ethyl Ester, N-[(5-Chloro-8-hydroxy-3-methyl-1-oxo-7-isochromanyl)carbonyl]-3-(phenyl-d5)-L-alanine Ethyl Ester, Ochratoxin A-d5 Ethyl Ester
InChI formel InChI=1S/C22H22ClNO6/c1-3-29-21(27)17(10-13-7-5-4-6-8-13)24-20(26)15-11-16(23)14-9-12(2)30-22(28)18(14)19(15)25/h4-8,11-12,17,25H,3,9-10H2,1-2H3,(H,24,26)/t12-,17+/m1/s1/i4D,5D,6D,7D,8D
SMIL [2H]c1c([2H])c([2H])c(C[C@H](NC(=O)c2cc(Cl)c3C[C@@H](C)OC(=O)c3c2O)C(=O)OCC)c([2H])c1[2H]
IUPAC navn ethyl (2S)-2-[[(3R)-5-chloro-8-hydroxy-3-methyl-1-oxo-3,4-dihydroisochromene-7-carbonyl]amino]-3-(2,3,4,5,6-pentadeuteriophenyl)propanoate
Molekylvægt (g/mol) 436.9
Formel vægt 436.1449