Learn More
Motic™ Objective, Plan Apochromat ULWD for PSM-1000
Ultra-long working distance objective suitable for PSM1000 Microscopes. Motic™ Objective, Objective, Plan Apochromat ULWD for PSM-1000 is designed for field curvature, spherical aberration, and chromatic aberration correction.
Brand:  Motic™ 1101001700082
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15485612
  • 20005.00 DKK / Each
Estimated Shipment
10-11-2020
Add to basket
Description
Description
  • Plan apochromat objective.
  • Ultra-long working distance.
  • Spherical aberration correction: 3-4 colors.
  • Chromatic aberration correction: 4-5 colors.
  • Field curvature correction.
Specifications
Specifications
WD: 13mm, NA: 0.550
Objective
100X
Plan apochromat, ultra-long working distance
PSM-1000 Probe Station Microscope