Learn More
Motic™ Objective, Plan Apochromat ELWD, Parfocality Adj. for PSM-1000
Extra-long working distance, parfocality adjustable objective suitable for PSM1000 Microscopes. Motic™ Objective, Plan Apochromat ELWD, Parfocality Adj. for PSM-1000 is designed for field curvature, spherical aberration, and chromatic aberration correction.
Brand:  Motic™ 1101001700141
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15485602
  • 5490.00 DKK / Each
Estimated Shipment
13-11-2020
Add to basket
Description
Description
  • Plan apochromat objective.
  • Extra-long working distance.
  • Parfocality adjustable.
  • Spherical aberration correction: 3-4 colors.
  • Chromatic aberration correction: 4-5 colors.
  • Field curvature correction.
Specifications
Specifications
WD: 34mm, NA: 0.055
Objective
2X
Plan apochromat, extra-long working distance, parfocality adjustable
PSM-1000 Probe Station Microscope