missing translation for 'onlineSavingsMsg'
Learn More

Motic™ Objective, Plan, Apochromat

Select a high-quality plan/apochromat objective for sharp, photo-quality images. Motic™ Objective, Plan, Apochromat is designed for field curvature, spherical aberration, and chromatic aberration correction.

5490.00 DKK - 19700.00 DKK

Specifications

Type Plan Apochromat
View More Specs

Products 4
Product Code Brand Description Magnification Power Price Quantity & Availability  
Product Code Brand Description Magnification Power Price Quantity & Availability  
15465292
View Documents Promotion Details
Motic™
1101001704731
NA: 0.35, WD: 4.2mm 10X This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15485292
View Documents Promotion Details
Motic™
1101001704751
NA: 0.95, WD: 0.18mm 40X This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15455292
View Documents Promotion Details
Motic™
1101001704721
NA: 0.15, WD: 20mm 4X This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15495292
View Documents Promotion Details
Motic™
1101001704821
NA: 1.3-Oil, WD: 0.25mm 100X This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Plan apochromat objective
  • Spherical aberration correction: 3-4 colors.
  • Chromatic aberration correction: 4-5 colors.
  • Field curvature correction.
Specifications

Specifications

Plan Apochromat
Videos
SDS
Documents

Documents

Product Certifications