Learn More
Motic™ Objective, Plan, Apochromat
Select a high-quality plan/apochromat objective for sharp, photo-quality images. Motic™ Objective, Plan, Apochromat is designed for field curvature, spherical aberration, and chromatic aberration correction.
Brand: Motic™ 1101001704821
Additional Details : Weight : 0.01000kg
Description
- Plan apochromat objective
- Spherical aberration correction: 3-4 colors.
- Chromatic aberration correction: 4-5 colors.
- Field curvature correction.
Specifications
Plan Apochromat | |
NA: 1.3-Oil, WD: 0.25mm | |
100X |