Learn More
Motic™ Objective, Plan, Apochromat
Select a high-quality plan/apochromat objective for sharp, photo-quality images. Motic™ Objective, Plan, Apochromat is designed for field curvature, spherical aberration, and chromatic aberration correction.
Brand:  Motic™ 1101001704751
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15485292
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Plan apochromat objective
  • Spherical aberration correction: 3-4 colors.
  • Chromatic aberration correction: 4-5 colors.
  • Field curvature correction.
Specifications
Specifications
Plan Apochromat
NA: 0.95, WD: 0.18mm
40X