Learn More
Motic™ Objective, CCIS Plan Achromatic EC-H PL for BA410
Color-corrected infinity system objective using multi-layer coated glass for enhanced contrast. Motic™ Objective, CCIS Plan Achromatic EC-H PL for BA410 is designed for field curvature, spherical aberration, and chromatic aberration correction.
Brand: Motic™ 1101001702941
Additional Details : Weight : 0.01000kg
Description
- Suitable for BA410 Series Upright Compound Microscope.
- Color-corrected infinity system.
- Enhanced contrast. Field curvature correction.
- Chromatic aberration correction for 2 colors.
- Spherical correction for 1 color.
Specifications
Objective | |
NA: 0.05, WD: 7.2mm | |
BA410E Microscopes | |
2X |