Learn More
Motic™ Objective, CCIS Plan Achromatic EC-H PL for BA410
Color-corrected infinity system objective using multi-layer coated glass for enhanced contrast. Motic™ Objective, CCIS Plan Achromatic EC-H PL for BA410 is designed for field curvature, spherical aberration, and chromatic aberration correction.
Brand:  Motic™ 1101001702871
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15485282
  • 675.00 DKK / Each
Estimated Shipment: 11-07-2024
to see stock.
Add to basket
Description
Description
  • Suitable for BA410 Series Upright Compound Microscope.
  • Color-corrected infinity system.
  • Enhanced contrast.
  • Field curvature correction.
  • Chromatic aberration correction for 2 colors.
  • Spherical correction for 1 color.
Specifications
Specifications
Objective
NA: 0.1, WD: 15.9mm
BA410E Microscopes
4X