missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OBFC1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99011DL594
Additional Details : Weight : 0.00970kg
Description
OBFC1 Polyclonal specifically detects OBFC1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
OBFC1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 594 | |
AAF44, AAF-44, alpha accessory factor 44, bA541N10.2, CST complex subunit STN1, oligonucleotide/oligosaccharide-binding fold containing 1, Oligonucleotide/oligosaccharide-binding fold-containing protein 1, replication protein A 32 kDa subunit, RP11-541N10.2, RPA-32, STN1FLJ22559, Suppressor of cdc thirteen homolog | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human OBFC1. | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
79991 | |
Store at 4°C in the dark. | |
IgG |