missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
OBFC1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99011AF647
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
OBFC1 Polyclonal specifically detects OBFC1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Tekniske data
| OBFC1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 647 | |
| AAF44, AAF-44, alpha accessory factor 44, bA541N10.2, CST complex subunit STN1, oligonucleotide/oligosaccharide-binding fold containing 1, Oligonucleotide/oligosaccharide-binding fold-containing protein 1, replication protein A 32 kDa subunit, RP11-541N10.2, RPA-32, STN1FLJ22559, Suppressor of cdc thirteen homolog | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human OBFC1. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 79991 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion