Få mere at vide
Tocris Bioscience™ OB-1
Beskrivelse
OB-1 is a stomatin-like protein-3 (STOML3) oligomerization inhibitor. Reduces STOML3 cluster size in the plasma membrane. Decreases mechanically-induced Piezo1 but not ASIC3 current amplitudes in N2a cells and cultured mouse sensory neurons in vitro. Subcutaneous administration in mice reversibly silences touch receptor activity and inhibits paw withdrawal reflex in a mouse neuropathic pain model.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl 5-(1-methyl-4-nitro-1H-imidazol-5-yloxy)-2-phenylbenzofuran-3-carboxylate |
| CAS | 300803-69-4 |
| Mængde | 50 mg |
| Molekylær formel | C21H17N3O6 |
| Renhed | 98% |