missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies O6-Benzylguanine

Artikelnummer. 17140272
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17140272 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17140272 Leverandør STEMCELL Technologies Leverandørnr. 73762

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Epigenetic modifier; Inactivates methylguanine DNA methyltransferase (MGMT)

  • O6-Benzylguanine is an efficient irreversible inactivator of the DNA repair protein O6-alkylguanine-DNA alkyltransferase (AGT, also known as methylguanine DNA methyltransferase, or MGMT)
  • AGT directly removes alkyl groups located on the O6-position of guanine from DNA, thereby restoring DNA integrity
  • O6-Benzylguanine is an antineoplastic agent that can be used to investigate the role of AGT in carcinogenesis and mutagenesis (Pegg 2011; Dolan et al.)
  • Enhances the activity of alkylating agents (nitrosourea, temozolomide, and cyclophosphamide) in malignant glioma xenografts growing in athymic nude mice (Pegg 1990)
  • Sensitizes CD34+ hematopoietic progenitors and a breast cancer cell line to bis-chloroethylnitrosourea (BCNU; Gerson et al.)

Tekniske data

CAS 19916-73-5
Synonym NSC 637037
Mængde 50 mg
Molekylær formel C12H11N5O
Renhed ≥98%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.