Få mere at vide
O5'-Succinylvitamin B12, TRC
Artikelnummer. 30425026
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30425026 10 mg
30410714 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30425026 Leverandør Toronto Research Chemicals Leverandørnr. TRCS69218010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale O5'-Succinylvitamin B12
CAS 55729-45-8
Analyt- eller komponentnavne O5'-Succinylvitamin B12
Forsendelsestilstand Room Temperature
Molekylær formel C67 H92 Co N 14 O17 P
Synonym Co-(cyano-κC)-cobinamide Dihydrogen Phosphate (Ester) Inner Salt, 3'-Ester with [1-[5-O-(3-Carboxy-1-oxopropyl)-α-D-ribofuranosyl]-5,6-dimethyl-1H-benzimidazole-κN3]
InChI formel InChI=1S/C66H94N13O17P.CN.Co/c1-31-22-41-42(23-32(31)2)79(30-74-41)61-56(90)57(43(94-61)29-93-53(89)19-18-52(87)88)96-97(91,92)95-33(3)28-73-51(86)20-21-63(8)39(24-48(70)83)60-66(11)65(10,27-50(72)85)38(14-17-47(69)82)55(78-66)35(5)59-64(9,26-49(71)84)36(12-15-45(67)80)40(75-59)25-44-62(6,7)37(13-16-46(68)81)54(76-44)34(4)58(63)77-60;1-2;/h22-23,25,30,33,36-39,43,56-57,60-61,90H,12-21,24,26-29H2,1-11H3,(H16,67,68,69,70,71,72,73,75,76,77,78,80,81,82,83,84,85,86,87,88,91,92);;/q;-1;+3/p-2
Løsningstype Neat
SMIL CC1CNC(=O)CCC2(C)C(CC(=O)N)C3C4(C)C(C)(CC(=O)N)C(CCC(=O)N)C5=N4[Co+3]67([C-]#N)(N8=CN(C9OC(COC(=O)CCC(=O)O)C(OP(=O)([O-])O1)C9O)c%10cc(C)c(C)cc8%10)N%11=C(C=C%12C(C)(C)C(CCC(=O)N)C(=N6%12)C(=C2[N-]37)C)C(CCC(=O)N)C(C)(CC(=O)N)C%11=C5C
Industry Type Forensics
Molekylvægt (g/mol) 1455.44
Mængde 10 mg
Formel vægt 1454.5834