Få mere at vide
Tocris Bioscience™ O4I2
GREENER_CHOICE
Oct3/4 inducer; induces expression of pluripotent-associated genes
Brand:  Tocris Bioscience™ 5664/10
Encompass
Produktkode 15737026
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
O4I2 is a Oct3/4 inducer; promotes Oct3/4 expression and stabilizes the Oct3/4 protein in HEK293 cells. Induces expression of pluripotent-associated genes Lin28, Sox2 and Nanog, and suppresses Rex1. Induces 15-fold increase in Oct3/4 expression in human primary fibroblasts.
Tekniske data
Tekniske data
Ethyl 2-[(4-chlorophenyl)amino]-4-thiazolecarboxylate
165682-93-9
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner