Få mere at vide
o-Tolunitrile-d7, CDN
Artikelnummer. 30313194
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313194 0.25 g
30318902 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313194 Leverandør CDN Leverandørnr. CDND63390.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methylbenzonitrile-D7
CAS 1138681-69-2
Molekylær formel C8 D7 N
Mængde 0.25 g
Synonym Benzonitrile-2,3,4,5-d4, 6-(methyl-d3)- (ACI), 6-(Methyl-d3)benzonitrile-2,3,4,5-d4 (ACI), o-Tolunitrile-d7
InChI formel InChI=1 S/C8H7N/c1-7-4-2-3-5-8(7)6-9/h2-5 H,1H3/i1D3,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c(c(C#N)c1[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2,3,4,5-tetradeuterio-6-(trideuteriomethyl)benzonitrile
Molekylvægt (g/mol) 124.191
Formel vægt 124.1018 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity