Få mere at vide
o-Toluidine-d9, CDN
Artikelnummer. 30315370
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30315370 1 g -
30313933 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315370 Leverandør CDN Leverandørnr. CDND35711G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Toluidine D9
CAS 194423-47-7
Molekylær formel C7 2H9 N
Mængde 1 g
Synonym Benzen-2,3,4,5-d4-amine-d2, 2-(methyl-d3)- (9 CI), 2-Toluidine-d9, o-Toluidine-d9, 2-Toluidine D9
InChI formel InChI=1 S/C7H9N/c1-6-4-2-3-5-7(6)8/h2-5 H,8H2,1H3/i1D3,2 D,3 D,4 D,5 D/hD2
SMIL [2 H]N([2 H])c1c([2 H])c([2 H])c([2 H])c([2 H])c1C([2 H])([2 H])[2 H]
IUPAC navn N,N,2,3,4,5-hexadeuterio-6-(trideuteriomethyl)aniline
Molekylvægt (g/mol) 116.21
Formel vægt 116.13 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity