Få mere at vide
o-Toluidine-d7 (NH2), CDN
Artikelnummer. 30311188
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311188 0.5 g
30315098 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311188 Leverandør CDN Leverandørnr. CDND78940.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Toluidine-d7
CAS 68408-22-0
Molekylær formel C7 D7 H2 N
Mængde 0.5 g
Synonym o-Toluidine-d7, 1-Amino-2-methylbenzene-d7, 2-Amino-1-methylbenzene-d7, 2-Aminotoluene-d7, 2-Methyl-1-aminobenzene-d7, 2-Methylaniline-d7, 2-Methylbenzenamine-d7, 2-Methylphenylamine-d7, 2-Tolylamine-d7, NSC 15348-d7, o-Aminotoluene-d7, o-Methylaniline-d7, o-Methylbenzenamine-d7;
InChI formel InChI=1 S/C7H9N/c1-6-4-2-3-5-7(6)8/h2-5 H,8H2,1H3/i1D3,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c(c(N)c1[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2,3,4,5-tetradeuterio-6-(trideuteriomethyl)aniline
Molekylvægt (g/mol) 114.196
Formel vægt 114.1174 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity