Få mere at vide
o-Toluidine-4,6-d2, CDN
Artikelnummer. 30317148
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30317148 0.5 g 0.5g
30327532 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317148 Leverandør CDN Leverandørnr. CDND56130.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale o-Toluidine-4,6-D2
CAS 68408-20-8
Molekylær formel C7 D2 H7 N
Mængde 0.5 g
Synonym Benzen-2,4-d2-amine, 6-methyl- (9 CI), 6-Methylbenzen-2,4-d2-amine, 4,6-Dideuterio-2-methyl-aniline, o-Toluidine-d2, o-Toluidine-4,6-D2, 2-Toluidine-4,6-d2
InChI formel InChI=1 S/C7H9N/c1-6-4-2-3-5-7(6)8/h2-5 H,8H2,1H3/i2D,5 D
SMIL [2 H]c1cc([2 H])c(N)c(C)c1
IUPAC navn 2,4-dideuterio-6-methylaniline
Molekylvægt (g/mol) 109.165
Formel vægt 109.086 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity