Få mere at vide
o-Toluic-D7 Acid, TRC
Artikelnummer. 30450094
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30450094 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30450094 Leverandør Toronto Research Chemicals Leverandørnr. TRCT536123250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale o-Toluic-D7 Acid
CAS 207742-73-2
Molekylær formel C8 D7 H O2
Mængde 250 mg
Synonym 6-(Methyl-D3)-benzoic-2,3,4,5-D4 Acid, Benzoic-2,3,4,5-d4 acid','6-(methyl-d3)- (9CI','ACI), 6-(Methyl-d3)benzoic-2,3,4,5-d4 acid (ACI), 2,3,4,5-Tetradeuterio-6-(trideuteriomethyl)benzoic acid, 2-Methylbenzoic acid D7
InChI formel InChI=1S/C8H8O2/c1-6-4-2-3-5-7(6)8(9)10/h2-5H,1H3,(H,9,10)/i1D3,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(c(C(=O)O)c1[2H])C([2H])([2H])[2H]
IUPAC navn 2,3,4,5-tetradeuterio-6-(trideuteriomethyl)benzoic acid
Molekylvægt (g/mol) 143.19
Formel vægt 143.0964