Få mere at vide
O-Tetrahydropyranyl Lubiprostone-d7, TRC
Artikelnummer. 30427683
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30427683 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427683 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2953825MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale O-Tetrahydropyranyl Lubiprostone-d7
CAS 1246812-24-7
Molekylær formel C25H33D7F2O6
Mængde 5 mg
Synonym (11alpha)-16,16-Difluoro-9,15-dioxo-11-[(tetrahydro-2H-pyran-2-yl)oxy]prostan-1-oic Acid-d7
InChI formel InChI=1S/C25H40F2O6/c1-2-3-15-25(26,27)22(29)14-13-19-18(10-6-4-5-7-11-23(30)31)20(28)17-21(19)33-24-12-8-9-16-32-24/h18-19,21,24H,2-17H2,1H3,(H,30,31)/t18-,19-,21-,24?/m1/s1/i1D3,2D2,3D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])CC(F)(F)C(=O)CC[C@@H]1[C@@H](CCCCCCC(=O)O)C(=O)C[C@H]1OC2CCCCO2
IUPAC navn 7-[(1R,2R,3R)-2-(6,6,7,7,8,8,8-heptadeuterio-4,4-difluoro-3-oxooctyl)-3-(oxan-2-yloxy)-5-oxocyclopentyl]heptanoic acid
Molekylvægt (g/mol) 481.62
Formel vægt 481.3232