missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ O-tert-Butyl-L-threonine, 97%

Brand:  Alfa Aesar™ H63031.03

Additional Details : CAS Number : 4378-13-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15479468

  • 218.52 DKK / 1g


Estimated Shipment
26-10-2020
Add to basket

Specifications

Chemical Identifiers

4378-13-6
175.228
NMJINEMBBQVPGY-RITPCOANSA-N
6951357
CC(C(C(=O)O)N)OC(C)(C)C
C8H17NO3
MFCD00037766
h-thr tbu-oh, o-tert-butyl-l-threonine, 2s,3r-2-amino-3-tert-butoxy butanoic acid, l-threonine, o-1,1-dimethylethyl, o-t-butyl-l-threonine, 2s,3r-2-amino-3-2-methylpropan-2-yl oxy butanoic acid, o-tert.-butylthreonin, o-tert.butyl-l-threonine, h-ser trt-2-chlorotrityl resin, h-thr tbu-2-chlorotrityl resin
(2S,3R)-2-amino-3-[(2-methylpropan-2-yl)oxy]butanoic acid

Specifications

O-tert-Butyl-L-threonine
C8H17NO3
2206127
NMJINEMBBQVPGY-RITPCOANSA-N
(2S,3R)-2-amino-3-[(2-methylpropan-2-yl)oxy]butanoic acid
6951357
Solid
1g
4378-13-6
MFCD00037766
h-thr tbu-oh, o-tert-butyl-l-threonine, 2s,3r-2-amino-3-tert-butoxy butanoic acid, l-threonine, o-1,1-dimethylethyl, o-t-butyl-l-threonine, 2s,3r-2-amino-3-2-methylpropan-2-yl oxy butanoic acid, o-tert.-butylthreonin, o-tert.butyl-l-threonine, h-ser trt-2-chlorotrityl resin, h-thr tbu-2-chlorotrityl resin
CC(C(C(=O)O)N)OC(C)(C)C
175.228
175.23
97%
Videos
SDS Documents