Learn More
Cytiva O-ring
Product Code. 11785185 Shop All Cytiva Products
Change view
Click to view available options
Size:
11.3 x 2.4mm
27.1 x 1.60 mm
37.47 x 5.34mm
6.75 x 1.78mm
Unit Size:
Each
Pack of 10
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11785185 11.3 x 2.4mm Each
10201640 37.47 x 5.34mm Pack of 5
11570854 27.1 x 1.60 mm Pack of 5
11530854 6.75 x 1.78mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11785185 Supplier Cytiva Supplier No. 18110497
  • ONLINE EXCLUSIVE PRICE
    2130.00 DKK
    1994.00 DKK / Pakke med 1
    Spare 136.00 DKK
    6% Slukket
På lager
Use in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules. Cytiva Life Sciences™ O-rings are made of nitrile rubber.
  • Can be used in aqueous solutions
  • Nearly all organic solvents
  • The following few exceptions: chlorinated hydrocarbons, acetone and otherketones, aliphatic esters and phenol. Solutions containingmore than 10% NaOH, 10% HCl, other strong mineral acidsor 5% acetic acid should not be used
  • Made of Nitrile Rubber (NBR)
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2/Pk.
Product Type O-Ring
Material Perfluor-Rubber
Size 11.3 x 2.4mm