Få mere at vide
o-Phthalaldehyde-d6, TRC
Artikelnummer. 30418020
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30418020 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418020 Leverandør Toronto Research Chemicals Leverandørnr. TRCP38420210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 68234-47-9
Molekylær formel C8D6O2
Molekylvægt (g/mol) 140.17
Synonym 1,2-Benzenedicarboxaldehyde-d6, 1,2-Diformylbenzene-d6, 1,2-Phthalaldehyde-d6, 2-Formylbenzaldehyde-d6, 2-Phthalaldehyde-d6, Benzene-1,2-dicarbaldehyde-d6, CIDEX OPA-d6, NSC 13394-d6, OP 100S-d6, OP 100SF-d6, Phthalic Aldehyde-d6, Phthalic Dialdehyde-d6, Phthalyldicarboxaldehyde-d6, Phtharal-d6, o-Benzenedicarbaldehyde-d6, o-Diformylbenzene-d6, Phthalaldehyde-d6, o-Phthaldialdehyde-d6, o-Phthalic Dialdehyde-d6
IUPAC navn o-Phthalaldehyde-d6
SMIL [2H]C(C1=C([2H])C([2H])=C([2H])C([2H])=C1C([2H])=O)=O
Tekniske data
Kemisk navn eller materiale o-Phthalaldehyde-d6
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C8H6O2/c9-5-7-3-1-2-4-8(7)6-10/h1-6H
Formel vægt 140.07
Procent renhed >95%