Få mere at vide
O-(Octadecylphosphoryl)choline, TRC
Artikelnummer. 30409942
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30409942 50 mg
30423643 250 mg
30413085 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30409942 Leverandør Toronto Research Chemicals Leverandørnr. TRCO59626050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 65956-63-0
Molekylær formel C23H50NO4P
Molekylvægt (g/mol) 435.63
Synonym Octadecylphosphorylcholine, Octadecylphosphocholine, NSC 282880, D 19391
IUPAC navn octadecyl 2-(trimethylazaniumyl)ethyl phosphate
SMIL CCCCCCCCCCCCCCCCCCOP(=O)([O-])OCC[N+](C)(C)C
Tekniske data
Kemisk navn eller materiale O-(Octadecylphosphoryl)choline
Mængde 50 mg
InChI formel InChI=1S/C23H50NO4P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-22-27-29(25,26)28-23-21-24(2,3)4/h5-23H2,1-4H3
Formel vægt 435.3477