missing translation for 'onlineSavingsMsg'
Få mere at vide

O,O-Diethyl Thiophosphate Potassium Salt, TRC

Artikelnummer. 30374726
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30374726 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30374726 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4452541G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale O,O-Diethyl hydrogen thiophosphate potassium salt
CAS 5871-17-0
Molekylær formel C4 H10 O3 P S . K
Mængde 1 g
Synonym Phosphorothioic acid, O,O-diethyl ester, potassium salt (1:1), Ethyl potassium phosphorothioate (6CI,7CI), Phosphorothioic acid, O,O-diethyl ester, potassium salt (8CI,9CI), O,O-Diethyl potassium phosphorothioate, Potassium O,O-diethyl phosphorothioate, Potassium O,O-diethylthiophosphate, Potassium di-O-ethyl phosphorothioate, Potassium diethyl thiophosphate
InChI formel InChI=1S/C4H11O3PS.K/c1-3-6-8(5,9)7-4-2;/h3-4H2,1-2H3,(H,5,9);/q;+1/p-1
SMIL [K+].CCOP(=O)([S-])OCC
IUPAC navn potassium;1-[ethoxy(sulfido)phosphoryl]oxyethane
Molekylvægt (g/mol) 208.26
Formel vægt 207.9725
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.